EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Cholinergic control of gamma power in the midbrain spatial attention network.
PMID 25589769
Abstract
The modulation of gamma power (25-90 Hz) is associated with attention and has been observed across species and brain areas. However, mechanisms that control these modulations are poorly understood. The midbrain spatial attention network in birds generates high-amplitude gamma oscillations in the local field potential that are thought to represent the highest priority location for attention. Here we explore, in midbrain slices from chickens, mechanisms that regulate the power of these oscillations, using high-resolution techniques including intracellular recordings from neurons targeted by calcium imaging. The results identify a specific subtype of neuron, expressing non-α7 nicotinic acetylcholine receptors, that directly drives inhibition in the gamma-generating circuit and switches the network into a primed state capable of producing high-amplitude oscillations. The special properties of this mechanism enable rapid, persistent changes in gamma power. The brain may employ this mechanism wherever rapid modulations of gamma power are critical to information processing.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1045009
Atropine sulfate, United States Pharmacopeia (USP) Reference Standard
C34H46N2O6 · H2O4S · H2O
A1400000
Atropine sulfate, European Pharmacopoeia (EP) Reference Standard
C34H46N2O6 · H2O4S · H2O
PHR1379
Atropine Sulfate, Pharmaceutical Secondary Standard; Certified Reference Material
C34H46N2O6 · H2O4S · H2O
11332
Atropine sulfate salt monohydrate, tested according to Ph.Eur.
C34H46N2O6 · H2O4S · H2O
A0257
Atropine sulfate salt monohydrate, ≥97% (TLC), crystalline
C34H46N2O6 · H2O4S · H2O
94563
Dimethyl sulfoxide, analytical standard
C2H6OS
01934
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
C2H6OS
1211006
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
C2H6OS
D2050000
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
C2H6OS
A5282
DL-2-Amino-5-phosphonopentanoic acid, solid
C5H12NO5P
54461
HEPES, ≥99.0% (T)
C8H18N2O4S
PHR1428
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
C8H18N2O4S
792519
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
CHNaO3
1613655
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
CHNaO3
71631
Sodium bicarbonate, tested according to Ph.Eur.
CHNaO3
84100
Sucrose, for microbiology, ACS reagent, ≥99.0%
C12H22O11
S1600000
Sucrose, European Pharmacopoeia (EP) Reference Standard
C12H22O11
1623637
Sucrose, United States Pharmacopeia (USP) Reference Standard
C12H22O11
PHR1001
Sucrose
C12H22O11
S1174
Sucrose, analytical standard, for enzymatic assay kit SCA20
C12H22O11