EMAIL THIS PAGE TO A FRIEND
Journal of visualized experiments : JoVE
Application of fluorescent nanoparticles to study remodeling of the endo-lysosomal system by intracellular bacteria.
PMID 25590656
Abstract
Fluorescent nanoparticles (NPs) with desirable chemical, optical and mechanical properties are promising tools to label intracellular organelles. Here, we introduce a method using gold-BSA-rhodamine NPs to label the endo-lysosomal system of eukaryotic cells and monitor manipulations of host cellular pathways by the intracellular pathogen Salmonella enterica. The NPs were readily internalized by HeLa cells and localized in late endosomes/lysosomes. Salmonella infection induced rearrangement of the vesicles and accumulation of NPs in Salmonella-induced membrane structures. We deployed the Imaris software package for quantitative analyses of confocal microscopy images. The number of objects and their size distribution in non-infected cells were distinct from the ones in Salmonella-infected cells, indicating extremely remodeling of the endo-lysosomal system by WT Salmonella.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
326542
Gold, beads, 1-6 mm, 99.999% trace metals basis
Au
373168
Gold, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Au
373176
Gold, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis
Au
373184
Gold, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Au
265829
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Au
349240
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Au
326496
Gold, foil, thickness 0.127 mm, 99.99% trace metals basis
Au
265810
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Au
349275
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Au
268461
Gold, foil, thickness 0.025 mm, 99.99% trace metals basis
Au
349267
Gold, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Au
255734
Gold, powder, <850 μm, ≥99.99% trace metals basis
Au
265772
Gold, powder, <45 μm, 99.99% trace metals basis
Au
718424
Gold, powder, size 10-20 μm
Au
326585
Gold, powder, <10 μm, ≥99.9% trace metals basis
Au
636347
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Au
265837
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Au
265802
Gold, wire, diam. 2.0 mm, 99.99% trace metals basis
Au
349291
Gold, wire, diam. 1.5 mm, 99.999% trace metals basis
Au
349305
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Au
265799
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis
Au
349313
Gold, wire, diam. 0.5 mm, 99.999% trace metals basis
Au
310980
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Au
265780
Gold, wire, diam. 0.25 mm, 99.99% trace metals basis
Au
326534
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Au
326526
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis
Au
349283
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Au
GF43139538
Gold, insulated wire, 5m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Au
GF92307922
Gold, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyester insulation, 99.99%
Au
GF11444611
Gold, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyimide insulation, 99.99%
Au