EMAIL THIS PAGE TO A FRIEND
Journal of visualized experiments : JoVE
Phosphopeptide analysis of rodent epididymal spermatozoa.
PMID 25591077
Abstract
Spermatozoa are quite unique amongst cell types. Although produced in the testis, both nuclear gene transcription and translation are switched off once the pre-cursor round cell begins to elongate and differentiate into what is morphologically recognized as a spermatozoon. However, the spermatozoon is very immature, having no ability for motility or egg recognition. Both of these events occur once the spermatozoa transit a secondary organ known as the epididymis. During the ~12 day passage that it takes for a sperm cell to pass through the epididymis, post-translational modifications of existing proteins play a pivotal role in the maturation of the cell. One major facet of such is protein phosphorylation. In order to characterize phosphorylation events taking place during sperm maturation, both pure sperm cell populations and pre-fractionation of phosphopeptides must be established. Using back flushing techniques, a method for the isolation of pure spermatozoa of high quality and yield from the distal or caudal epididymides is outlined. The steps for solubilization, digestion, and pre-fractionation of sperm phosphopeptides through TiO2 affinity chromatography are explained. Once isolated, phosphopeptides can be injected into MS to identify both protein phosphorylation events on specific amino acid residues and quantify the levels of phosphorylation taking place during the sperm maturation processes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
IRMM531A
Titanium, IRMM® certified Reference Material, 0.1 mm foil
Ti
IRMM531B
Titanium, IRMM® certified Reference Material, 0.5 mm foil
Ti
IRMM531C
Titanium, IRMM® certified Reference Material, 0.5 mm wire
Ti
305812
Titanium, crystalline, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Ti
433667
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Ti
369489
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Ti
348805
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Ti
267481
Titanium, foil, thickness 0.25 mm, 99.99% trace metals basis
Ti
267503
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Ti
460397
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Ti
348791
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Ti
348813
Titanium, foil, thickness 0.1 mm, 99.99% trace metals basis
Ti
348821
Titanium, foil, thickness 0.05 mm, 99.99% trace metals basis
Ti
348848
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Ti
268496
Titanium, powder, −100 mesh, 99.7% trace metals basis
Ti
366994
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Ti
513415
Titanium, nanoparticles, dispersion, <100 nm particle size, in mineral oil, 98.5% trace metals basis
Ti
347132
Titanium, rod, diam. 6.35 mm, 99.99% trace metals basis
Ti
266051
Titanium, rod, diam. 6.35 mm, 99.7% trace metals basis
Ti
268526
Titanium, sponge, 3-19 mm, 99.5% trace metals basis
Ti
348856
Titanium, wire, diam. 2.0 mm, 99.99% trace metals basis
Ti
266035
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Ti
267902
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Ti
348864
Titanium, wire, diam. 0.5 mm, 99.99% trace metals basis
Ti
460400
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Ti
266019
Titanium, wire, diam. 0.127 mm, 99.99% trace metals basis
Ti
749044
Titanium, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.995% trace metals basis
Ti
767506
Titanium, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.995% trace metals basis
Ti
GF57414179
Titanium, rod, 1000mm, diameter 6mm, annealed, 99.6+%
Ti
GF72693883
Titanium, rod, 100mm, diameter 1.5mm, annealed, 99.6+%
Ti