EMAIL THIS PAGE TO A FRIEND
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Expansion of myeloid-derived suppressor cells in patients with acute coronary syndrome.
PMID 25591771
Abstract
The aim of this study was to explore whether the circulating frequency and function of myeloid-derived suppressor cells (MDSCs) are altered in patients with acute coronary syndrome (ACS). The frequency of MDSCs in peripheral blood was determined by flow cytometry, and mRNA expression in purified MDSCs was analyzed by real-time reverse transcription polymerase chain reaction (RT-PCR). The suppressive function of MDSCs isolated from different groups was also determined. The plasma levels of certain cytokines were determined using Bio-Plex Pro™ Human Cytokine Assays. The frequency of circulating CD14(+)HLA-DR(-/low) MDSCs; arginase-1 (Arg-1) expression; and plasma levels of interleukin (IL)-1β, IL-6, tumor necrosis factor (TNF)-α, and IL-33 were markedly increased in ACS patients compared to stable angina (SA) or control patients. Furthermore, MDSCs from ACS patients were more potent suppressors of T-cell proliferation and IFN-γ production than those from the SA or control groups at ratios of 1:4 and 1:2; this effect was partially mediated by Arg-1. In addition, the frequency of MDSCs was positively correlated with plasma levels of IL-6, IL-33, and TNF-α. We observed an increased frequency and suppressive function of MDSCs in ACS patients, a result that may provide insights into the mechanisms involved in ACS.