EMAIL THIS PAGE TO A FRIEND
Nature
Blown-up brains reveal nanoscale details.
PMID 25592513
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
408220
Sodium acrylate, 97%
C3H3NaO2
409464
Zinc acrylate, 98%
C6H6O4Zn