EMAIL THIS PAGE TO A FRIEND
EMBO molecular medicine
An aberrant sugar modification of BACE1 blocks its lysosomal targeting in Alzheimer's disease.
PMID 25592972
Abstract
The β-site amyloid precursor protein cleaving enzyme-1 (BACE1), an essential protease for the generation of amyloid-β (Aβ) peptide, is a major drug target for Alzheimer's disease (AD). However, there is a concern that inhibiting BACE1 could also affect several physiological functions. Here, we show that BACE1 is modified with bisecting N-acetylglucosamine (GlcNAc), a sugar modification highly expressed in brain, and demonstrate that AD patients have higher levels of bisecting GlcNAc on BACE1. Analysis of knockout mice lacking the biosynthetic enzyme for bisecting GlcNAc, GnT-III (Mgat3), revealed that cleavage of Aβ-precursor protein (APP) by BACE1 is reduced in these mice, resulting in a decrease in Aβ plaques and improved cognitive function. The lack of this modification directs BACE1 to late endosomes/lysosomes where it is less colocalized with APP, leading to accelerated lysosomal degradation. Notably, other BACE1 substrates, CHL1 and contactin-2, are normally cleaved in GnT-III-deficient mice, suggesting that the effect of bisecting GlcNAc on BACE1 is selective to APP. Considering that GnT-III-deficient mice remain healthy, GnT-III may be a novel and promising drug target for AD therapeutics.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31573
5α-Androstan-17β-ol-3-one, VETRANAL, analytical standard
C19H30O2
71251
Acetic acid, analytical standard
C2H4O2
MAB374
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, 200 µg, Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5 is a well published and extensively characterized monoclonal antibody. This purified mAb detects Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH) & has been published & validated for use in ELISA, IP, IC, IF, IH & WB.
MAB386
Anti-Myelin Basic Protein Antibody, a.a. 82-87, 1 mL, Anti-Myelin Basic Protein Antibody, a.a. 82-87 is an antibody against Myelin Basic Protein for use in ELISA, IC, IH, RIA & WB.
90577
Bifido Selective Supplement B, for microbiology
C2H4O2
C3350000
Cytarabine, European Pharmacopoeia (EP) Reference Standard
C9H13N3O5
1162002
Cytarabine, United States Pharmacopeia (USP) Reference Standard
C9H13N3O5
C6645
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
C9H13N3O5 · HCl
1005706
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
C2H4O2
791741
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
C2H3NaO2
S2889
Sodium acetate, anhydrous, for molecular biology, ≥99%
C2H3NaO2
S3272
Sodium acetate, for electrophoresis, ≥99%, crystalline
C2H3NaO2
S5636
Sodium acetate, powder, BioReagent, for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
C2H3NaO2
S1429
Sodium acetate, meets USP testing specifications, anhydrous
C2H3NaO2
1613407
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
C2H3NaO2
229873
Sodium acetate, 99.995% trace metals basis
C2H3NaO2
S7545
Sodium acetate, BioXtra, ≥99.0%
C2H3NaO2
110191
Sodium acetate, ReagentPlus®, 99%
C2H3NaO2
71183
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
C2H3NaO2
W302406
Sodium Acetate Anhydrous, >99%, FCC, FG
C2H3NaO2
S8388
Sodium acetate solution, 0.3 M
C2H3NaO2
71196
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
C2H3NaO2
613053
Sodium acetate solution, NMR reference standard, 50 mM in D2O (99.9 atom % D), water 1 %, NMR tube size 3 mm × 8 in.
C2H3NaO2
Y0000620
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
C12H25NaO4S