EMAIL THIS PAGE TO A FRIEND
Indian journal of pharmaceutical sciences
A Facile Synthesis, In vitro Antiinflammatory and Antioxidant activity of Novel Benzimidazolylpyrano[2,3-d][1,3]thiazolocarbonitriles.
PMID 25593384
Abstract
The synthesis benzimidazolylpyrano [2,3-d] [1,3] thiazolocarbonitriles (5a-j) were achieved by cyclocondensation of arylidene amino-benzo[d]imidazole-2-thiols (3a-j) with mercaptoacetic acid followed by cyclization with 2-(phenylmethylene)malononitrile. Further more, the present study aimed at the evaluation of in vitro antiinflammatory activity and antioxidant activity of synthetic compounds. All tested compounds showed appreciable activity against the standard drugs.