EMAIL THIS PAGE TO A FRIEND
Environmental pollution (Barking, Essex : 1987)
Effect of transpiration on plant accumulation and translocation of PPCP/EDCs.
PMID 25594843
Abstract
The reuse of treated wastewater for agricultural irrigation in arid and hot climates where plant transpiration is high may affect plant accumulation of pharmaceutical and personal care products (PPCPs) and endocrine disrupting chemicals (EDCs). In this study, carrot, lettuce, and tomato plants were grown in solution containing 16 PPCP/EDCs in either a cool-humid or a warm-dry environment. Leaf bioconcentration factors (BCF) were positively correlated with transpiration for chemical groups of different ionized states (p < 0.05). However, root BCFs were correlated with transpiration only for neutral PPCP/EDCs (p < 0.05). Neutral and cationic PPCP/EDCs showed similar accumulation, while anionic PPCP/EDCs had significantly higher accumulation in roots and significantly lower accumulation in leaves (p < 0.05). Results show that plant transpiration may play a significant role in the uptake and translocation of PPCP/EDCs, which may have a pronounced effect in arid and hot climates where irrigation with treated wastewater is common.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284785
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
C14H14O3
197777
2-(p-Chlorophenoxy)-2-methylpropionic acid, 97%
C10H11ClO3
94496
Carbamazepine, analytical standard
C15H12N2O
C0450000
Carbamazepine, European Pharmacopoeia (EP) Reference Standard
C15H12N2O
1093001
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
C15H12N2O
PHR1067
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
C15H12N2O
90323
Clofibric acid, analytical reference material
C10H11ClO3
D2425
Diuron, ≥98%
C9H10Cl2N2O
45463
Diuron, PESTANAL®, analytical standard
C9H10Cl2N2O
G9518
Gemfibrozil
C15H22O3
91823
Gemfibrozil, analytical standard
C15H22O3
Y0000513
Gemfibrozil, European Pharmacopoeia (EP) Reference Standard
C15H22O3
PHR1286
Gemfibrozil, Pharmaceutical Secondary Standard; Certified Reference Material
C15H22O3
Y0000514
Gemfibrozil for system suitability, European Pharmacopoeia (EP) Reference Standard
C15H22O3
M2392
Meprobamate, analytical standard
C9H18N2O4
M0400000
Meprobamate, European Pharmacopoeia (EP) Reference Standard
C9H18N2O4
N8280
Naproxen, meets USP testing specifications
C14H14O3
46482
Naproxen, VETRANAL, analytical standard
C14H14O3
PHR1040
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
C14H14O3
N0250000
Naproxen, European Pharmacopoeia (EP) Reference Standard
C14H14O3
1457301
Naproxen, United States Pharmacopeia (USP) Reference Standard
C14H14O3
33824
Pentadecafluorooctanoic acid, analytical standard
C8HF15O2
33603
Pentadecafluorooctanoic acid solution, 100 μg/mL in methanol, analytical standard
C8HF15O2
171468
Perfluorooctanoic acid, 96%
C8HF15O2
P7295
Primidone, analytical standard
C12H14N2O2
P2950000
Primidone, European Pharmacopoeia (EP) Reference Standard
C12H14N2O2
Y0000369
Primidone for peak identification, European Pharmacopoeia (EP) Reference Standard
C12H14N2O2
S7507
Sulfamethoxazole, analytical standard
C10H11N3O3S
76177
Sulfamethoxazole, certified reference material, TraceCERT®
C10H11N3O3S
31737
Sulfamethoxazole, VETRANAL, analytical standard
C10H11N3O3S