EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
First characterisation of flavonoid- and diarylheptanoid-type antioxidant phenolics in Corylus maxima by HPLC-DAD-ESI-MS.
PMID 25594894
Abstract
Corylus maxima Mill. (Betulaceae) leaves have been used in traditional medicine both internally and externally, nevertheless phytochemical exploration of the plant remains incomplete. In this study, the in vitro antioxidant activity and polyphenolic composition of the ethyl acetate and methanolic extracts of C. maxima leaves and bark are reported for the first time. The radical scavenging activities of the extracts were investigated by the ABTS and DPPH assays. All the extracts of C. maxima possessed notable antioxidant activity. By mean of a HPLC-DAD-ESI-TOF and a HPLC-DAD-ESI-MS/MS method, altogether twenty-two phenolics were tentatively characterised: one flavan derivative (1), seven flavonol derivatives (4, 6, 12, 13, 16, 20 and 21) and fourteen diarylheptanoids (2, 3, 5, 7-11, 14, 15, 17-19 and 22). The amount of the two main flavonoids - myricetin-3-O-rhamnoside (6) and quercetin-3-O-rhamnoside (13) - and two diarylheptanoids - oregonin (3) and hirsutenone (15) - in the extracts were determined by a validated HPLC-ESI-MS/MS method in multiple reaction monitoring (MRM) mode. Our results showed that C. maxima could be considered as a valuable source of pharmacologically important natural products that might contribute to the revaluation of the phytotherapeutical potential of the plant.
Related Materials