EMAIL THIS PAGE TO A FRIEND
Nuclear medicine and biology
Automated synthesis and PET evaluation of both enantiomers of [¹⁸F]FMISO.
PMID 25595134
Abstract
[(18)F]FMISO, the widely used positron emission tomography (PET) hypoxia tracer, is a chiral compound clinically used as a racemic mixture. The purpose of this study was to synthesize the individual (R)- and the (S)- enantiomers of [(18)F]FMISO and compare their PET imaging characteristics. The radiosynthesis of enantiopure (R)- and (S)-[(18)F]FMISO was based on Co(salen) (N,N'-bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt)-mediated opening of enantiopure epoxides with [(18)F]HF. The uptake and clearance of the individual [(18)F]FMISO antipodes were investigated using micro-PET/CT imaging performed on mice bearing FaDu tumors. Image-derived biodistribution was obtained from micro-PET/CT scans performed at 1 and 3 hours post injection (p.i.). In addition, the uptake patterns of each enantiomer were observed using two-hour dynamic micro-PET/CT scans, and the time-activity curves from different organs were compared. The individual (R)- and (S)-[(18)F]FMISO enantiomers were synthesized in one step with high enantiomeric excess (ee)>99% and radiochemical purity>97% using custom-made automation module. The dynamic micro-PET/CT scanning revealed a faster initial uptake of the (R)-[(18)F]FMISO enantiomer in tumor and muscle tissues, however the difference became progressively smaller with time. The tumor-to-muscle (T/M) and tumor-to-liver (T/L) ratios remained nearly identical for the (R)- and (S)-forms at all time points. The micro-PET/CT imaging at 1 and 3 hours p.i. did not show any significant enantioselective tissue uptake. Although the (R)-enantiomer of [(18)F]FMISO demonstrated a somewhat faster initial tumor and muscle uptake no significant enantioselective tissue uptake was observed at later time points. The T/M- and T/L- ratios for the (R)- and (S)-forms were the same within the experimental error at all times. Therefore, the use of enantiopure [(18)F]FMISO is unlikely to present any practical clinical benefit for PET imaging.