EMAIL THIS PAGE TO A FRIEND
Circulation. Cardiovascular imaging
Relative flow reserve derived from quantitative perfusion imaging may not outperform stress myocardial blood flow for identification of hemodynamically significant coronary artery disease.
PMID 25596142
Abstract
Quantitative myocardial perfusion imaging is increasingly used for the diagnosis of coronary artery disease. Quantitative perfusion imaging allows to noninvasively calculate fractional flow reserve (FFR). This so-called relative flow reserve (RFR) is defined as the ratio of hyperemic myocardial blood flow (MBF) in a stenotic area to hyperemic MBF in a normal perfused area. The aim of this study was to assess the value of RFR in the detection of significant coronary artery disease. From a clinical population of patients with suspected coronary artery disease who underwent oxygen-15-labeled water cardiac positron emission tomography and invasive coronary angiography, 92 patients with single- or 2-vessel disease were included. Intermediate lesions (diameter stenosis, 30%-90%; n=75) were interrogated by FFR. Thirty-eight (41%) vessels were deemed hemodynamically significant (>90% stenosis or FFR≤0.80). Hyperemic MBF, coronary flow reserve, and RFR were lower for vessels with a hemodynamically significant lesion (2.01±0.78 versus 2.90±1.16 mL·min(-1)·g(-1); P<0.001, 2.27±1.03 versus 3.10±1.29; P<0.001, and 0.67±0.23 versus 0.93±0.15; P<0.001, respectively). The correlation between RFR and FFR was moderate (r=0.54; P<0.01). Receiver operator characteristic curve analysis showed an area under the curve of 0.82 for RFR, which was not significantly higher compared with that for hyperemic MBF and coronary flow reserve (0.76; P=0.32 and 0.72; P=0.08, respectively). Noninvasive estimation of FFR by quantitative perfusion positron emission tomography by calculating RFR is feasible, yet only a trend toward a slight improvement of diagnostic accuracy compared with hyperemic MBF assessment was determined.