EMAIL THIS PAGE TO A FRIEND
Ophthalmology
Bowman layer transplantation to reduce and stabilize progressive, advanced keratoconus.
PMID 25596620
Abstract
To evaluate the clinical outcome of mid-stromal isolated Bowman layer transplantation, a new surgical technique to reduce and stabilize ectasia in eyes with advanced keratoconus, to postpone penetrating keratoplasty or deep anterior lamellar keratoplasty, and to enable continued daily contact lens wear. Prospective, nonrandomized cohort study at a tertiary referral center. Twenty-two eyes of 19 patients with progressive, advanced keratoconus not eligible for ultraviolet cross-linking. The mid-stroma was manually dissected and an isolated donor Bowman layer was positioned within the stromal pocket. Before and up to 36 months after surgery (mean follow-up, 21±7 months), best spectacle-corrected visual acuity (BSCVA), best contact lens-corrected visual acuity (BCLVA), Scheimpflug-based corneal tomography measurements, endothelial cell density, biomicroscopy, refraction, and intraoperative and postoperative complications were recorded. Two surgeries were complicated by an intraoperative perforation of Descemet membrane; no other intraoperative or postoperative complications were observed. Maximum keratometry decreased on average from 77.2±6.2 diopters (D) to 69.2±3.7 D (P < 0.001) at 1 month after surgery and remained stable thereafter (P ≥ 0.072). Mean BSCVA improved from 1.27±0.44 logarithm of the minimum angle of resolution units before surgery to 0.90±0.30 logarithm of the minimum angle of resolution units 12 months after surgery (P < 0.001), whereas BCLVA remained stable (P = 0.105). Mean thinnest-point pachymetry increased from 332±59 μm before surgery to 360±50 μm at the latest follow-up (P = 0.012), and no change in endothelial cell density was found (P = 0.355). With isolated Bowman layer transplantation, reduction and stabilization of corneal ectasia was achieved in eyes with progressive, advanced keratoconus. Given the low risk for complications, the procedure may be performed to postpone penetrating or deep anterior lamellar keratoplasty.