EMAIL THIS PAGE TO A FRIEND
Animal science journal = Nihon chikusan Gakkaiho
Zinc-bearing zeolite clinoptilolite improves tissue zinc accumulation in laying hens by enhancing zinc transporter gene mRNA abundance.
PMID 25597922
Abstract
A study was conducted to investigate effects of zinc-bearing zeolite clinoptilolite (ZnCP), as an alternative for zinc sulfate (ZnSO4), on laying performance, tissue Zn accumulation and Zn transporter genes expression in laying hens. Hy-Line Brown laying hens were allocated to three treatments, each of which had six replicates with 15 hens per replicate, receiving basal diet supplemented with ZnSO4 (control, 80 mg Zn/kg diet), 0.23% ZnCP (40.25 mg Zn/kg diet) and 0.46% ZnCP (80.50 mg Zn/kg diet) for 8 weeks, respectively. Compared with control, hens fed diet containing 0.23% ZnCP had similar Zn content in measured tissues (P > 0.05). A higher ZnCP inclusion (0.46%) enhanced Zn accumulation in liver (P < 0.05) and pancreas (P < 0.05). In addition, ZnCP inclusion increased blood iron (Fe) content (P < 0.05). ZnCP supplementation enhanced jejunal metallothionein-4 (MT-4) messenger RNA (mRNA) abundance (P < 0.05). ZnCP inclusion at a higher level (0.46%) increased mRNA expression of MT-4 in pancreas (P < 0.05) and zinc transporter-1 (ZnT-1) in jejunum (P < 0.05). The highest ZnT-2 mRNA abundance in jejunum was found in hens fed 0.23% ZnCP inclusion diet (P < 0.05). The results indicated that ZnCP reached a higher bioavailability as compared with ZnSO4 as evidenced by enhanced tissue Zn accumulation and Zn transporter genes expression.