EMAIL THIS PAGE TO A FRIEND
Letters in applied microbiology
Highly cold-active pectinases under wine-like conditions from non-Saccharomyces yeasts for enzymatic production during winemaking.
PMID 25598190
Abstract
The influence of oenological factors on cold-active pectinases from 15 preselected indigenous yeasts belonging to Aureobasidium pullulans, Filobasidium capsuligenum, Rhodotorula dairenensis, Cryptococcus saitoi and Saccharomyces cerevisiae was investigated. Pectinolytic enzymes were constitutive or partially constitutive; and high glucose concentration (200xa0gxa0l(-1) ) did not affect or increased pectinase production at 12°C and pH 3·5 (up to 113·9xa0Uxa0mg(-1) ) only in A.xa0pullulans strains. SO2 (120xa0mgxa0l(-1) ) slightly affected the growth of A.xa0pullulans strains but did not affect pectinase production levels. Ethanol (15%) barely affected pectinase activity of A.xa0pullulans strains but diminished relative activity to 12-79% of basidiomycetous yeasts. Moreover, non-Saccharomyces strains showed promising properties of oenological interest. This study demonstrates that cold-active pectinases from some A.xa0pullulans strains were able to remain active at glucose, ethanol and SO2 concentrations usually found in vinification, and suggests their potential use as processing aids for low-temperature winemaking. Nowadays, there is increasing interest in low-temperature winemaking. Nevertheless, commercial oenological pectinases, produced by fungi, are rarely active at low temperatures. Cold-active pectinases that are stable under vinification conditions are needed. This study indicated that cold-active and acid-tolerant pectinases from non-Saccharomcyes yeasts were able to remain active at glucose, ethanol and SO2 concentrations usually found in winemaking. Furthermore, not only are these yeasts a source of cold-active pectinases, but the yeasts themselves are also potential adjunct cultures for oenology to produce these enzymes during cold-winemaking.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5879
O-(4-Nitrophenylphosphoryl)choline
C11H17N2O6P