EMAIL THIS PAGE TO A FRIEND
Neuroscience research
Evaluation for roles of neurosteroids in modulating forebrain mechanisms controlling vasopressin secretion and related phenomena in conscious rats.
PMID 25598212
Abstract
Anteroventral third ventricular region (AV3V) that regulates autonomic functions through a GABAergic mechanism possesses neuroactive steroid (NS)-synthesizing ability. Although NS can exert effects by acting on a certain type of GABAA-receptor (R), it is not clear whether NS may operate to modulate AV3V GABAergic activity for controlling autonomic functions. This study aimed to investigate the issue. AV3V infusion with a GABAA antagonist bicuculline increased plasma vasopressin (AVP), glucose, blood pressure (BP), and heart rate in rats. These events were abolished by preinjecting its agonist muscimol, whereas the infusion with allopregnanolone, a NS capable of potentiating GABAA-R function, affected none of the variables in the absence or presence of such bicuculline actions. Similarly, AV3V infusion with pregnanolone sulfate, a NS capable of antagonizing GABAA-R, produced no effect on those variables. AV3V infusion with muscimol was effective in inhibiting the responses of plasma AVP or glucose, or BP to an osmotic loading or bleeding. However, AV3V infusion with aminoglutethimide, a NS synthesis inhibitor, did not affect any of the variables in the absence or presence of those stimuli. These results suggest that NS may not cause acute effects on the AV3V GABAergic mechanism involved in regulating AVP release and other autonomic function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P8887
5α-Pregnan-3α-ol-20-one, solid
C21H34O2
P162
Pregnenolone sulfate sodium salt
C21H31NaO5S