EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. C, Nanomaterials and interfaces
Metal-Enhanced Fluorescence from Silver Nanowires with High Aspect Ratio on Glass Slides for Biosensing Applications.
PMID 25598859
Abstract
High enhancement of fluorescence emission, improved fluorophore photostability, and significant reduction of fluorescence lifetimes have been obtained from high aspect ratio (>100) silver (Ag) nanowires. These quantities are found to depend on the surface loading of Ag nanowires on glass slides, where the enhancement of fluorescence emission increases with the density of nanowires. The surface loading dependence was attributed to the creation of intense electric fields around the network of Ag nanowires and to the coupling of fluorophore excited states that takes place efficiently at a distance of 10 nm from the surface of nanowires, which was confirmed by theoretical calculations. The enhancement of fluorescence emission of fluorescein isothiocyanate (FITC) was assessed by fluorescence spectroscopy and fluorescence-lifetime imaging microscopy (FLIM) to demonstrate the potential of high aspect ratio Ag nanowires. Fluorescence enhancement factors exceeding 14 were observed on Ag nanowires with high loading by FLIM. The photostability of FITC was the highest on nanowires with medium loading under continuous laser excitation for 10 min because of the significant reduction in the fluorescence lifetime of FITC on these surfaces. These results clearly demonstrate the potential of Ag nanowires in metal-enhanced fluorescence-based applications of biosensing on planar surfaces and cellular imaging.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
808644 Silver, Nanowires, diam. × L 60±5 nm × 40±5 μm, 5 mg/mL in ETH
806609 Silver, nanowires, diam. × L 20 nm (±2 nm) × 12 μm (±2 μm), 5 mg/mL (in water)
806617 Silver, Nanowires, diam. × L 20 nm (±2 nm) × 12 μm (±2 μm), 5 mg/mL (in ETH)
806714 Silver, Nanowires, diam. × L 20 nm (±2 nm) × 12 μm (±2 μm), 5 mg/mL (in IPA)
806722 Silver, Nanowires, diam. × L 25 nm (±3 nm) × 15 μm (±2 μm), 5 mg/mL (in water)
806838 Silver, Nanowires, diam. × L 25 nm (±3 nm) × 15 μm (±2 μm), 5 mg/mL (in ETH)
806846 Silver, Nanowires, diam. × L 25 nm (±3 nm) × 15 μm (±2 μm), 5 mg/mL (in IPA)
806935 Silver, Nanowires, diam. × L 30 nm (±5 nm) × 20 μm (±3 μm), 5 mg/mL (in water)
806943 Silver, Nanowires, diam. × L 30 nm (±5 nm) × 20 μm (±3 μm), 5 mg/mL (in ETH)
807052 Silver, Nanowires, diam. × L 30 nm (±5 nm) × 20 μm (±3 μm), 5 mg/mL (in IPA)
807060 Silver, Nanowires, diam. × L 35 nm (±5 nm) × 25 μm (±5 μm), 5 mg/mL (in water)
807168 Silver, Nanowires, diam. × L 35 nm (±5 nm) × 25 μm (±5 μm), 5 mg/mL (in ETH)
807176 Silver, Nanowires, diam. × L 35 nm (±5 nm) × 25 μm (±5 μm), 5 mg/mL (in IPA)
807265 Silver, Nanowires, diam. × L 40 nm (±5 nm) × 35 μm (±5 μm), 5 mg/mL (in water)
807273 Silver, Nanowires, diam. × L 40 nm (±5 nm) × 35 μm (±5 μm), 5 mg/mL (in ETH)
807389 Silver, Nanowires, diam. × L 40 nm (±5 nm) × 35 μm (±5 μm), 5 mg/mL (in IPA)
807397 Silver, Nanowires, diam. × L 50 nm (±10 nm) × 40 μm (±5 μm), 5 mg/mL (in water)
807494 Silver, Nanowires, diam. × L 50 nm (±10 nm) × 40 μm (±5 μm), 5 mg/mL (in ETH)
807508 Silver, Nanowires, diam. × L 50 nm (±10 nm) × 40 μm (±5 μm), 5 mg/mL (in IPA)
807591 Silver, Nanowires, diam. × L 60 nm (±10 nm) × 40 μm (±5 μm), 5 mg/mL (in water)
807702 Silver, Nanowires, diam. × L 60 nm (±10 nm) × 40 μm (±5 μm), 5 mg/mL (in IPA)
807826 Silver, Nanowires, diam. × L 70 nm (±10 nm) × 40 μm (±5 μm), 5 mg/mL (in water)
807923 Silver, Nanowires, diam. × L 70 nm (±10 nm) × 40 μm (±5 μm), 5 mg/mL (in ETH)
808032 Silver, Nanowires, diam. × L 70 nm (±10 nm) × 40 μm (±5 μm), 5 mg/mL (in IPA)