EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Simultaneous determination of insulin and its analogues in pharmaceutical formulations by micellar electrokinetic chromatography.
PMID 25599910
Abstract
A simple and efficient MEKC method was developed to simultaneously determine human insulin, its five analogues, the main degradation products and the excipients usually present in injection formulations. A very fast method with a total analysis time of 3 min was then successfully validated for the analysis of human insulin and the quality control of commercial formulations was carried out.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W353000
m-Cresol, ≥98%, FG
C7H8O