EMAIL THIS PAGE TO A FRIEND
Digestive diseases and sciences
Regional Bias of Intratumoral Genetic Heterogeneity of Apoptosis-Related Genes BAX, APAF1, and FLASH in Colon Cancers with High Microsatellite Instability.
PMID 25599959
Abstract
Apoptosis inactivation and intratumoral heterogeneity (ITH) are common features of cancers, including colorectal cancer (CRC). Inactivation of apoptosis prolongs cancer cell survival, and ITH may contribute to CRC progression. To examine the presence and extent of mutational ITH in the pro-apoptotic genes APAF1, BAX, and FLASH and the association of mutational ITH with pathologic parameters of CRC. The ITH of mutations in the mononucleotide repeats of APAF1, BAX and FLASH in different tumors were analyzed in 16 cases of CRC with high microsatellite instability (MSI-H) and 41 cases of CRC with stable MSI/low MSI (MSS/MSI-L) by single-strand conformation polymorphism and DNA sequencing analyses. Frameshift mutations of APAF1, BAX, and FLASH were identified in 19, 31, and 6 % of CRC with MSI-H, respectively, but also in cases of CRC with MSS/MSI-L. All but one CRC with a mutation (8/9) harbored regional ITH of the APAF1, BAX and FLASH frameshift mutations. ITH, however, was not associated with histopathologic features of CRC with MSI-H, suggesting that ITH might not be related to development of the MSI-H phenotype itself, but rather to disease progression. Our results indicate that the APAF1, BAX, and FLASH genes not only harbor frameshift mutations but also demonstrate mutational ITH, which together might play a role in the tumorigenesis of CRC with MSI-H by affecting the apoptosis of cancer cells. Our data also suggest that multiregional mutation analysis is needed for a better evaluation of the mutation status in CRC.