EMAIL THIS PAGE TO A FRIEND
BMC developmental biology
Development of the follicular basement membrane during human gametogenesis and early folliculogenesis.
PMID 25605128
Abstract
In society, there is a clear need to improve the success rate of techniques to restore fertility. Therefore a deeper knowledge of the dynamics of the complex molecular environment that regulates human gametogenesis and (early) folliculogenesis in vivo is necessary. Here, we have studied these processes focusing on the formation of the follicular basement membrane (BM) in vivo. The distribution of the main components of the extracellular matrix (ECM) collagen IV, laminin and fibronectin by week 10 of gestation (W10) in the ovarian cortex revealed the existence of ovarian cords and of a distinct mesenchymal compartment, resembling the organization in the male gonads. By W17, the first primordial follicles were assembled individually in that (cortical) mesenchymal compartment and were already encapsulated by a BM of collagen IV and laminin, but not fibronectin. In adults, in the primary and secondary follicles, collagen IV, laminin and to a lesser extent fibronectin were prominent in the follicular BM. The ECM-molecular niche compartimentalizes the female gonads from the time of germ cell colonization until adulthood. This knowledge may contribute to improve methods to recreate the environment needed for successful folliculogenesis in vitro and that would benefit a large number of infertility patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8417
4′,6-Diamidino-2-phenylindole dihydrochloride, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC), suitable for fluorescence
C16H15N5 · 2HCl
32670
4′,6-Diamidino-2-phenylindole dihydrochloride, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)
C16H15N5 · 2HCl
AB748
Anti-Collagen Type IV Antibody, Anti-Collagen Type IV Antibody detects level of Collagen Type IV & has been published & validated for use in ELISA, IH.
02672
Calcium atomic spectroscopy standard concentrate 1.00 g Ca, 1.00 g/L, for 1L standard solution, analytical standard
CaCl2
21059
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
CaCl2
21049
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
CaCl2
1012772
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
C2H6O
PHR1070
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
C2H6O
95660
o-Xylene, analytical standard
C8H10
295884
o-Xylene, for HPLC, 98%
C8H10
PHR1312
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10
T1378
Sigma 7-9®, ≥99% (titration), crystalline
C4H11NO3
154563
Tris(hydroxymethyl)aminomethane, ultrapure grade, ≥99.9%
C4H11NO3
252859
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
C4H11NO3
RDD008
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
C4H11NO3
T1503
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
C4H11NO3
T6066
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, cell culture tested, ≥99.9% (titration)
C4H11NO3
T6791
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
C4H11NO3
T4661
Trizma® base, ≥99.9% (titration), crystalline
C4H11NO3
93362
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
C4H11NO3
93352
Trizma® base, ≥99.0% (T)
C4H11NO3
93350
Trizma® base, puriss. p.a., ≥99.7% (T)
C4H11NO3
33742
Trizma® base, puriss. p.a., buffer substance, ≥99.5%
C4H11NO3
04577
Trizma® base, tested according to Ph.Eur.
C4H11NO3
93440
Trizma® base, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
C4H11NO3
T2550000
Trometamol, European Pharmacopoeia (EP) Reference Standard
C4H11NO3
T6687
Tromethamine, meets USP testing specifications
C4H11NO3
RES3137T-A7
Tromethamine, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production., suitable for cell culture, meets USP testing specifications
C4H11NO3
RES3193T-A7
Tromethamine, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture, Meets USP, EP, JPC, BP testing specifications.
C4H11NO3
PHR1347
Tromethamine, Pharmaceutical Secondary Standard; Certified Reference Material
C4H11NO3