EMAIL THIS PAGE TO A FRIEND
The Journal of physiology
The use of murine-derived fundic organoids in studies of gastric physiology.
PMID 25605613
Abstract
An in vitro approach to study gastric development is primary mouse-derived epithelium cultured as three-dimensional spheroids known as organoids. We have devised two unique gastric fundic-derived organoid cultures: model 1 for the expansion of gastric fundic stem cells, and model 2 for the maintenance of mature cell lineages. Organoids maintained in co-culture with immortalized stomach mesenchymal cells express robust numbers of surface pit, mucous neck, chief, endocrine and parietal cells. Histamine induced a significant decrease in intraluminal pH that was reversed by omeprazole in fundic organoids and indicated functional activity and regulation of parietal cells. Localized photodamage resulted in rapid cell exfoliation coincident with migration of neighbouring cells to the damaged area, sustaining epithelial continuity. We report the use of these models for studies of epithelial cell biology and cell damage and repair. Studies of gastric function and disease have been limited by the lack of extended primary cultures of the epithelium. An in vitro approach to study gastric development is primary mouse-derived antral epithelium cultured as three-dimensional spheroids known as organoids. There have been no reports on the use of organoids for gastric function. We have devised two unique gastric fundic-derived organoid cultures: model 1 for the expansion of gastric fundic stem cells, and model 2 for the maintenance of mature cell lineages. Both models were generated from single glands dissociated from whole fundic tissue and grown in basement membrane matrix (Matrigel) and organoid growth medium. Model 1 enriches for a stem cell-like niche via simple passage of the organoids. Maintained in Matrigel and growth medium, proliferating organoids expressed high levels of stem cell markers CD44 and Lgr5. Model 2 is a system of gastric organoids co-cultured with immortalized stomach mesenchymal cells (ISMCs). Organoids maintained in co-culture with ISMCs express robust numbers of surface pit, mucous neck, chief, endocrine and parietal cells. Histamine induced a significant decrease in intraluminal pH that was reversed by omeprazole in fundic organoids and indicated functional activity and regulation of parietal cells. Localized photodamage resulted in rapid cell exfoliation coincident with migration of neighbouring cells to the damaged area, sustaining epithelial continuity. Thus, we report the use of these models for studies of epithelial cell biology and cell damage and repair.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1300015
Anti-DCAMKL1 (C-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1305446
ANTI-DCAMKL1(C-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1305341
ANTI-DCAMKL1(N-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4200294
Anti-DCLK1 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB4200186
Anti-DCLK1 antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody
HPA015655
Anti-DCLK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA039908
Anti-GIF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA040774
Anti-GIF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410525
Anti-GIF antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
O104
Omeprazole, solid
C17H19N3O3S
19329
Omeprazole, analytical reference material
C17H19N3O3S
PHR1059
Omeprazole, Pharmaceutical Secondary Standard; Certified Reference Material
C17H19N3O3S
1478505
Omeprazole, United States Pharmacopeia (USP) Reference Standard
C17H19N3O3S
O0150000
Omeprazole, European Pharmacopoeia (EP) Reference Standard
C17H19N3O3S
Y0001042
Omeprazole for peak identification, European Pharmacopoeia (EP) Reference Standard
C17H19N3O3S