EMAIL THIS PAGE TO A FRIEND
Autophagy
Poly-ADP-ribosylation of HMGB1 regulates TNFSF10/TRAIL resistance through autophagy.
PMID 25607248
Abstract
Both apoptosis ("self-killing") and autophagy ("self-eating") are evolutionarily conserved processes, and their crosstalk influences anticancer drug sensitivity and cell death. However, the underlying mechanism remains unclear. Here, we demonstrated that HMGB1 (high mobility group box 1), normally a nuclear protein, is a crucial regulator of TNFSF10/TRAIL (tumor necrosis factor [ligand] superfamily, member 10)-induced cancer cell death. Activation of PARP1 (poly [ADP-ribose] polymerase 1) was required for TNFSF10-induced ADP-ribosylation of HMGB1 in cancer cells. Moreover, pharmacological inhibition of PARP1 activity or knockdown of PARP1 gene expression significantly inhibited TNFSF10-induced HMGB1 cytoplasmic translocation and subsequent HMGB1-BECN1 complex formation. Furthermore, suppression of the PARP1-HMGB1 pathway diminished autophagy, increased apoptosis, and enhanced the anticancer activity of TNFSF10 in vitro and in a subcutaneous tumor model. These results indicate that PARP1 acts as a prominent upstream regulator of HMGB1-mediated autophagy and maintains a homeostatic balance between apoptosis and autophagy, which provides new insight into the mechanism of TNFSF10 resistance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2502092
Anti-FBL antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502093
Anti-FBL antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2701663
Anti-fbl antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701789
Anti-fbl antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV40366
Anti-FBL antibody produced in rabbit, IgG fraction of antiserum
SAB4300633
Anti-Fibrillarin antibody produced in rabbit, affinity isolated antibody
EHU072991 MISSION® esiRNA, esiRNA human HMGB1 (esiRNA1)
EHU024471 MISSION® esiRNA, esiRNA human FADD (esiRNA1)
EHU050101 MISSION® esiRNA, esiRNA human PARP1 (esiRNA1)
EHU152091 MISSION® esiRNA, esiRNA human CASP8 (esiRNA1)
EHU061741 MISSION® esiRNA, esiRNA human BECN1 (esiRNA1)
SHC001
MISSION® pLKO.1-puro Empty Vector Control Plasmid DNA, Contains no shRNA insert
WH0002091M1
Monoclonal Anti-FBL antibody produced in mouse, clone 4G9-E4, purified immunoglobulin, buffered aqueous solution
SAB1403793
Monoclonal Anti-FBL antibody produced in mouse, clone 3E10, purified immunoglobulin, buffered aqueous solution
N9037
Necrostatin-1, ≥98% (HPLC)
C13H13N3OS
P5493 Phosphate buffered saline, 10× concentrate, BioPerformance Certified, suitable for cell culture
P4365
PJ-34 hydrochloride hydrate, ≥98% (HPLC), powder
C17H17N3O2 · HCl · xH2O
T9701 TRAIL human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE and HPLC), lyophilized powder
P1379
TWEEN® 20, viscous liquid
C1230
Z-Ile-Glu(O-ME)-Thr-Asp(O-Me) fluoromethyl ketone, ≥90% (TLC), powder
C30H43FN4O11