EMAIL THIS PAGE TO A FRIEND
Experimental and clinical endocrinology & diabetes : official journal, German Society of Endocrinology [and] German Diabetes Association
The comparative effect of pioglitazone and metformin on serum osteoprotegerin, adiponectin and intercellular adhesion molecule concentrations in patients with newly diagnosed type 2 diabetes: a randomized clinical trial.
PMID 25607338
Abstract
The etiologic role of inflammatory pathways in the development of diabetic complications, especially cardiovascular events, has been established. The anti-inflammatory role of metformin and pioglitazone has been described; however, no study to date has compared the efficacy of these common oral agents in this regard. In this study, the authors aimed to compare the anti-inflammatory properties of pioglitazone and metformin, with respect to their effect on serum concentrations of highly sensitive C-reactive protein (hsCRP), osteoprotegerin (OPG), intercellular adhesion molecule-1 (ICAM-1) and adiponectin. In an open-label randomized clinical trial, 117 patients with newly diagnosed type 2 diabetes mellitus were visited; 84 fulfilled the inclusion criteria, and were randomly allocated to 2 arms receiving either 1,000 mg/d metformin or 30 mg/d pioglitazone, respectively. Biochemical assessments were made at baseline and the end of the 3 months trial. Significant reduction in FPG, insulin and HbA1c in women and men of both arms were observed. Log-hsCRP values significantly decreased in both arms. A decreasing, but non-significant trend in log-OPG levels was observed in women of the metformin arm (p=0.063). A greater reduction in log-ICAM levels was identifiable in men receiving pioglitazone compared to the other arm (p=0.008); in addition, the same trend was observed in log-OPG values (p=0.029). Nonetheless, reduction in log-ICAM and log-OPG levels was comparable between the 2 arms. A significant increase in adiponectin was observed in both men and women in the pioglitazone arm (p<0.001), whereas changes were non-significant in the metformin arm. Remarkably, patients receiving pioglitazone revealed more significant reduction in inflammatory markers.