EMAIL THIS PAGE TO A FRIEND
Nature communications
Whole-exome sequencing identifies somatic ATRX mutations in pheochromocytomas and paragangliomas.
PMID 25608029
Abstract
Pheochromocytomas and paragangliomas (PCC/PGL) are the solid tumour type most commonly associated with an inherited susceptibility syndrome. However, very little is known about the somatic genetic changes leading to tumorigenesis or malignant transformation. Here we perform whole-exome sequencing on a discovery set of 21 PCC/PGL and identify somatic ATRX mutations in two SDHB-associated tumours. Targeted sequencing of a separate validation set of 103 PCC/PGL identifies somatic ATRX mutations in 12.6% of PCC/PGL. PCC/PGL with somatic ATRX mutations are associated with alternative lengthening of telomeres and clinically aggressive behaviour. This finding suggests that loss of ATRX, an SWI/SNF chromatin remodelling protein, is important in the development of clinically aggressive PCC/PGL.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA001906
Anti-ATRX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA008736
Anti-DAXX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
F9037
Formamide, ≥99.5% (GC), BioReagent, for molecular biology
CH3NO
F7508
Formamide, for molecular biology, ≥99.5% (GC), liquid
CH3NO
221198
Formamide, ACS reagent, ≥99.5%
CH3NO
47671
Formamide, BioUltra, for molecular biology, ≥99.5% (T)
CH3NO
185906
Formamide, reagent grade, 98%
CH3NO
613339
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
CH3NO
551384
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
CH3NO
612707
Formamide solution, NMR reference standard, 45% in DMSO-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
CH3NO
611891
Formamide solution, NMR reference standard, 45% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
CH3NO
733784
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
CH3NO
736201
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
CH3NO