EMAIL THIS PAGE TO A FRIEND
Molecular plant-microbe interactions : MPMI
Ethylene Signaling Modulates Herbivore-Induced Defense Responses in the Model Legume Medicago truncatula.
PMID 25608182
Abstract
One or more effectors in the labial saliva (LS) of generalist Noctuid caterpillars activate plant signaling pathways to modulate jasmonate (JA)-dependent defense responses; however, the exact mechanisms involved have yet to be elucidated. A potential candidate in this phytohormone interplay is the ethylene (ET) signaling pathway. We compared the biochemical and molecular responses of the model legume Medicago truncatula and the ET-insensitive skl mutant to herbivory by fourth instar Spodoptera exigua (Hübner) caterpillars with intact or impaired LS secretions. Cellular oxidative stress increases rapidly after herbivory, as evidenced by changes in oxidized-to-reduced ascorbate (ASC) and glutathione (GSH) ratios. The caterpillar-specific increase in GSH ratios and the LS-specific increase in ASC ratios are alleviated in the skl mutant, indicating that ET signaling is required. Ten hours postherbivory, markers of the JA and JA/ET pathways are differentially expressed; MtVSP is induced and MtHEL is repressed in a caterpillar LS- and ET-independent manner. In contrast, expression of the classic marker of the systemic acquired resistance pathway, MtPR1, is caterpillar LS-dependent and requires ET signaling. Caterpillar LS further suppresses the induction of JA-related trypsin inhibitor activity in an ET-dependent manner. Findings suggest that ET is involved in the caterpillar LS-dependent, salicylic acid/NPR1-mediated attenuation of JA-dependent induced responses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
536164
Ethylene, 99.99%
C2H4
295329
Ethylene, ≥99.5%
C2H4