EMAIL THIS PAGE TO A FRIEND
CNS neuroscience & therapeutics
Surgical stress induces brain-derived neurotrophic factor reduction and postoperative cognitive dysfunction via glucocorticoid receptor phosphorylation in aged mice.
PMID 25611431
Abstract
This study explored whether surgical stress-induced glucocorticoid receptor (GR) phosphorylation is related to postoperative cognitive dysfunction (POCD) in aged individuals. Inhibition of GR activation could be an effective treatment for POCD. A laparotomy was given to C57/BL6 mice in POCD group both 20 and 6 months old. Animals in control group were treated in identical manners except for laparotomy. Cognitive function was evaluated by Morris water maze and elevated plus maze. Western blot and Elisa assay were used to detect related molecules. Mifepristone and roscovitine were treated as inhibitions of GR phosphorylation. The cognitive function was impaired, and brain-derived neurotrophic factor (BDNF) was found reduced in aged POCD group. GR translocation into nucleus and elevated GR phosphorylation were found in prefrontal cortex of aged POCD mice. Cyclin-dependent Kinase 5 (CDK5), kinase for GR phosphorylation also elevated in aged POCD mice. With GR antagonist and CDK5 inhibitor, reduction of BDNF and cognitive dysfunction in aged mice were both rescued. These results presented a mechanism that surgical stress-induced GR phosphorylation contributes to POCD in aged individuals. Inhibition of GR activation and phosphorylation might be a potential treatment target of POCD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
55417
1,2-Propanediol, analytical reference material
C3H8O2
12279
1,2-Propanediol, analytical standard
C3H8O2
82281
1,2-Propanediol, tested according to Ph.Eur.
C3H8O2
HPA011271
Anti-ANXA1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA011272
Anti-ANXA1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV36568
Anti-ANXA1 antibody produced in rabbit, IgG fraction of antiserum
HPA023691
Anti-CDCA5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100385
Anti-CDCA5 antibody produced in rabbit, affinity isolated antibody
SAB2106191
Anti-CDCA5 antibody produced in rabbit, affinity isolated antibody
AV38525
Anti-CDK5 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV38526
Anti-CDK5 (AB2) antibody produced in rabbit, IgG fraction of antiserum
HPA018977
Anti-CDK5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100390
Anti-CDK5 antibody produced in rabbit, affinity isolated antibody
SAB2106207
Anti-CDK5R1 antibody produced in rabbit, affinity isolated antibody
SAB2700744
Anti-GCGR (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2100906
Anti-GCHFR antibody produced in rabbit, affinity isolated antibody
SAB4501139
Anti-GCHFR antibody produced in rabbit, affinity isolated antibody
HPA046258
Anti-GCHFR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410517
Anti-GCHFR antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4501309
Anti-GR antibody produced in rabbit, affinity isolated antibody
SAB4501310
Anti-GR antibody produced in rabbit, affinity isolated antibody
SAB4501311
Anti-GR antibody produced in rabbit, affinity isolated antibody
HPA001886 Anti-IL12A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA000701
Anti-MNAT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV100992
Anti-MNAT1 antibody produced in rabbit, affinity isolated antibody
AV38150
Anti-MNAT1 antibody produced in rabbit, IgG fraction of antiserum
HPA004248
Anti-NR3C1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101649
Anti-NR3C1 antibody produced in rabbit, affinity isolated antibody
SAB1408583
Anti-NR3C1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4504585
Anti-phospho-GR (pSer203) antibody produced in rabbit, affinity isolated antibody