EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Thymoquinone induces caspase-independent, autophagic cell death in CPT-11-resistant lovo colon cancer via mitochondrial dysfunction and activation of JNK and p38.
PMID 25611974
Abstract
Chemotherapy causes unwanted side effects and chemoresistance, limiting its effectiveness. Therefore, phytochemicals are now used as alternative treatments. Thymoquinone (TQ) is used to treat different cancers, including colon cancer. The irinotecan-resistant (CPT-11-R) LoVo colon cancer cell line was previously constructed by stepwise CPT-11 challenges to untreated parental LoVo cells. TQ dose-dependently increased the total cell death index and activated apoptosis at 2 μM, which then diminished at increasing doses. The possibility of autophagic cell death was then investigated. TQ caused mitochondrial outer membrane permeability (MOMP) and activated autophagic cell death. JNK and p38 inhibitors (SP600125 and SB203580, respectively) reversed TQ autophagic cell death. TQ was also found to activate apoptosis before autophagy, and the direction of cell death was switched toward autophagic cell death at initiation of autophagosome formation. Therefore, TQ resulted in caspase-independent, autophagic cell death via MOMP and activation of JNK and p38 in CPT-11-R LoVo colon cancer cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
274666
Thymoquinone, ≥98%
C10H12O2