EMAIL THIS PAGE TO A FRIEND
Molecular cancer therapeutics
ATF4 Gene Network Mediates Cellular Response to the Anticancer PAD Inhibitor YW3-56 in Triple-Negative Breast Cancer Cells.
PMID 25612620
Abstract
We previously reported that a pan-PAD inhibitor, YW3-56, activates p53 target genes to inhibit cancer growth. However, the p53-independent anticancer activity and molecular mechanisms of YW3-56 remain largely elusive. Here, gene expression analyses found that ATF4 target genes involved in endoplasmic reticulum (ER) stress response were activated by YW3-56. Depletion of ATF4 greatly attenuated YW3-56-mediated activation of the mTORC1 regulatory genes SESN2 and DDIT4. Using the ChIP-exo method, high-resolution genomic binding sites of ATF4 and CEBPB responsive to YW3-56 treatment were generated. In human breast cancer cells, YW3-56-mediated cell death features mitochondria depletion and autophagy perturbation. Moreover, YW3-56 treatment effectively inhibits the growth of triple-negative breast cancer xenograft tumors in nude mice. Taken together, we unveiled the anticancer mechanisms and therapeutic potentials of the pan-PAD inhibitor YW3-56.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3500210
Anti-4E-BP1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500736
Anti-4E-BP1 antibody produced in rabbit, affinity isolated antibody
SAB4500737
Anti-4E-BP1 antibody produced in rabbit, affinity isolated antibody
AV38067
Anti-ATF4 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2501899
Anti-ATF4 (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2500130
Anti-ATF4 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2100168
Anti-ATF4 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2105294
Anti-ATF4, (C-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4500112
Anti-C/EBP-β antibody produced in rabbit, affinity isolated antibody
SAB1405605
Anti-CEBPB antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV31259
Anti-CEBPB antibody produced in rabbit, affinity isolated antibody
SAB2100397
Anti-CEBPB antibody produced in rabbit, affinity isolated antibody
SAB4500113
Anti-CEBPB, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2701263
Anti-DCTN4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2107220
Anti-DCTN4 antibody produced in rabbit, affinity isolated antibody
SAB2105017
Anti-DCTN4, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1405706
Anti-DDIT3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1300754
Anti-DDIT4 (C-term) antibody produced in rabbit, Ig fraction of antiserum, buffered aqueous solution
SAB1300087
Anti-DDIT4 (N-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1407555
Anti-DDIT4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA034508
Anti-DDIT4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300474
Anti-EIF4EBP1 (Ab-36) antibody produced in rabbit, affinity isolated antibody
SAB4300475
Anti-EIF4EBP1 (Ab-45) antibody produced in rabbit, affinity isolated antibody
HPA023501
Anti-EIF4EBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106955
Anti-EIF4EBP1 antibody produced in rabbit, affinity isolated antibody
G6916 Anti-GADD153 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500632
Anti-GADD153 antibody produced in rabbit, affinity isolated antibody
GW22005A Anti-GTF2H1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW22005B Anti-GTF2H1 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV100811
Anti-GTF2H1 antibody produced in rabbit, IgG fraction of antiserum