EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
PEGylated D-serine dehydratase as a D-serine reducing agent.
PMID 25617179
Abstract
D-Serine is an endogenous coagonist for N-methyl-D-aspartate (NMDA) receptors and is involved in excitatory neurotransmission. Excessive receptor activation causes excitotoxicity, leading to various acute and chronic neurological disorders. Decrease in D-serine content may provide a therapeutic strategy for the treatment of the neurological disorders in which overstimulation of NMDA receptors plays a pathological role. Saccharomyces cerevisiaed-serine dehydratase (Dsd1p), which acts dominantly on D-serine, may be a useful D-serine reducing agent. We conjugated a linear 5-kDa polyethylene glycol (PEG) to Dsd1p (PEG-Dsd1p) and examined the effects of PEG-conjugation on its biochemical and pharmacokinetic properties. PEG-Dsd1p retained activity, specificity, and stability of the enzyme. The PEG modification extended the serum half-life of Dsd1p in mice 6-fold, from 3.8h to 22.4h. PEG-Dsd1p was much less immunogenic compared to the unmodified enzyme. Intraperitoneal administration of PEG-Dsd1p was effective in decreasing the D-serine content in the mouse hippocampus. These findings suggest that PEG-Dsd1p may be a novel tool for lowering D-serine levels in vivo.