EMAIL THIS PAGE TO A FRIEND
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS)
A study of lipid- and water-soluble arsenic species in liver of Northeast Arctic cod (Gadus morhua) containing high levels of total arsenic.
PMID 25618262
Abstract
In the present study liver samples (n=26) of Northeast Arctic cod (Gadus morhua), ranging in total arsenic concentrations from 2.1 to 240mg/kg liver wet weight (ww), were analysed for their content of total arsenic and arsenic species in the lipid-soluble and water-soluble fractions. The arsenic concentrations in the lipid fractions ranged from 1.8 to 16.4mg As/kg oil of liver, and a linear correlation (r(2)=0.80, p<0.001) was observed between the total arsenic concentrations in liver and the total arsenic concentrations in the respective lipid fractions of the same livers. The relative proportion of arsenolipids was considerably lower in liver samples with high total arsenic levels (33-240mg/kg ww), which contained from 3 to 7% of the total arsenic in the lipid-soluble fraction. In contrast liver samples with low arsenic concentrations (2.1-33mg/kg ww) contained up to 50% of the total arsenic as lipid-soluble species. Arsenic speciation analysis of the lipid-soluble fractions of the livers, using reversed-phase high performance liquid chromatography coupled to inductively coupled plasma mass spectrometry (HPLC-ICP-MS), revealed the presence of several arsenolipids. Three major arsenic-containing hydrocarbons (C17H39AsO, C19H41AsO and C23H37AsO) and five arsenic-containing fatty acids (C17H35AsO3, C19H39AO3, C19H37AsO3, C23H37AsO3 and C24H37AsO3) were identified using HPLC coupled to quadrupole time-of-flight mass spectrometry (qTOF-MS). Arsenobetaine was the major arsenic species in the water-soluble fraction of the livers, while dimethylarsinate, arsenocholine and inorganic arsenic were minor constituents. Inorganic arsenic accounted for less than 0.1% of the total arsenic in the liver samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS013577
ammonium carbonate, AldrichCPR
C2H13N3O6
379999
Ammonium carbonate, 99.999% trace metals basis
CH8N2O3
207861
Ammonium carbonate, ACS reagent, ≥30.0% NH3 basis
CH8N2O3
11204
Ammonium carbonate, puriss., meets analytical specification of NF, Ph. Franc., FCC
CH8N2O3
00752
Argon, ≥99.998%
Ar
202657
Arsenic, 99.999% trace metals basis
As
267961
Arsenic, powder, ≥99.997% trace metals basis
As
239240
Cesium, ingot, ≥99.95% trace metals basis
Cs
494526
Heptane, biotech. grade, ≥99%
C7H16
154873
Heptane, spectrophotometric grade, 99%
C7H16
51730
Heptane, analytical standard
C7H16
PHR1554
Heptane, Pharmaceutical Secondary Standard; Certified Reference Material
C7H16
277932
Thallium, granular, 1-5 mm diameter, 99.99% trace metals basis
Tl
GF28981941
Thallium, rod, 95mm, diameter 10.0mm, extruded, 99.99+%
Tl
GF50522228
Thallium, rod, 198mm, diameter 10.0mm, extruded, 99.99+%
Tl
GF74984473
Thallium, rod, 250mm, diameter 8.0mm, extruded, 99.99+%
Tl
T81906
Triphenylarsine, 97%
C18H15As
262994
Yttrium, chips, 99.9% trace rare earth metals basis
Y
451347
Yttrium, dendritic pieces, 99.9% trace metals basis
Y
693642
Yttrium, foil, thickness 1 mm, 99.8% trace rare earth metals basis
Y
261319
Yttrium, ingot, 99.9% trace rare earth metals basis
Y
261327
Yttrium, powder, −40 mesh, 99.5% trace rare earth metals basis
Y
GF09119279
Yttrium, foil, 25x25mm, thickness 0.15mm, as rolled, 99%
Y
GF77674719
Yttrium, wire reel, 0.2m, diameter 0.5mm, 99.9%
Y
GF11301398
Yttrium, rod, 50mm, diameter 6.35mm, cast, 99%
Y
GF86394156
Yttrium, wire reel, 0.2m, diameter 1.0mm, 99.9%
Y
GF91377478
Yttrium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Y
GF87149481
Yttrium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Y
GF03037148
Yttrium, foil, 50x50mm, thickness 0.15mm, as rolled, 99%
Y
GF55118257
Yttrium, foil, 25mm disks, thickness 0.50mm, as rolled, 99%
Y