EMAIL THIS PAGE TO A FRIEND
Journal of environmental health
A study on exposure to cyanide among a community living near a gold mine in Malaysia.
PMID 25619035
Abstract
Cyanidation in gold extraction has detrimental impacts on the ecosystem and the population in surrounding areas. The research described in this article aimed to assess the health effects of environmental exposure to cyanide in a community near a gold mining area in Malaysia. A total of 255 members of the exposed community and 117 members of the control community answered questionnaires on their exposure to a gold mine and health outcomes. Urine samples were collected for biomarker assessment (i.e., urinary thiocyanate). The authors found that the mean level of urinary thiocyanate was significantly higher among the exposed community (0.30 ± 0.26 mg/dL) than the control community (0.24 ± 0.23 mg/dL) (p = .0021). The exposed community had a significantly higher prevalence of headaches (prevalence odds ratio [pOR] = 2.6, p = .001), dizziness (pOR = 3.7, p < .001), skin irritation (pOR = 2.1, p = .040), and eye irritation (pOR = 5.1, p = .004). No association occurred between dietary exposure and urinary biomarkers. Smokers living near a gold mine had a higher urinary thiocyanate level (0.56 mg/dL) than smokers in the control community (0.49 mg/dL).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
326542
Gold, beads, 1-6 mm, 99.999% trace metals basis
Au
373168
Gold, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Au
373176
Gold, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis
Au
373184
Gold, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Au
265829
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Au
349240
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Au
326496
Gold, foil, thickness 0.127 mm, 99.99% trace metals basis
Au
265810
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Au
349275
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Au
268461
Gold, foil, thickness 0.025 mm, 99.99% trace metals basis
Au
349267
Gold, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Au
255734
Gold, powder, <850 μm, ≥99.99% trace metals basis
Au
265772
Gold, powder, <45 μm, 99.99% trace metals basis
Au
718424
Gold, powder, size 10-20 μm
Au
326585
Gold, powder, <10 μm, ≥99.9% trace metals basis
Au
636347
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Au
265837
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Au
265802
Gold, wire, diam. 2.0 mm, 99.99% trace metals basis
Au
349291
Gold, wire, diam. 1.5 mm, 99.999% trace metals basis
Au
349305
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Au
265799
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis
Au
349313
Gold, wire, diam. 0.5 mm, 99.999% trace metals basis
Au
310980
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Au
265780
Gold, wire, diam. 0.25 mm, 99.99% trace metals basis
Au
326534
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Au
326526
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis
Au
349283
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Au
GF43139538
Gold, insulated wire, 5m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Au
GF92307922
Gold, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyester insulation, 99.99%
Au
GF11444611
Gold, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyimide insulation, 99.99%
Au