EMAIL THIS PAGE TO A FRIEND
Pesticide biochemistry and physiology
A novel Pro197Glu substitution in acetolactate synthase (ALS) confers broad-spectrum resistance across ALS inhibitors.
PMID 25619909
Abstract
Water chickweed (Myosoton aquaticum L.), a competitive broadleaf weed, is widespread in wheat fields in China. Tribenuron and pyroxsulam failed to control water chickweed in the same field in Qiaotian Village in 2011 and 2012, respectively. An initial tribenuron resistance confirmation test identified a resistant population (AH02). ALS gene sequencing revealed a previously unreported substitution of Glu for Pro at amino acid position 197 in resistant individuals. A purified subpopulation (WRR04) that was individually homozygous for the Pro197Glu substitution was generated and characterized in terms of its response to different classes of ALS inhibitors. A whole-plant experiment showed that the WRR04 population exhibited broad-spectrum resistance to tribenuron (SU, 318-fold), pyrithiobac sodium (PTB, > 197-fold), pyroxsulam (TP, 81-fold), florasulam (TP, > 36-fold) and imazethapyr (IMI, 11-fold). An in vitro ALS assay confirmed that the ALS from WRR04 showed high resistance to all the tested ALS inhibitors. These results established that the Pro197Glu substitution endows broad-spectrum resistance across ALS inhibitors in water chickweed. In addition, molecular markers were developed to rapidly identify the Pro197Glu mutation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
37923
Imazethapyr, PESTANAL®, analytical standard
C15H19N3O3