EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Major pathways of polymyxin-induced apoptosis in rat kidney proximal tubular cells.
PMID 25624331
Abstract
Identifying the pathways involved in the apoptotic cell death that is associated with polymyxin-induced nephrotoxicity is crucial for the development of strategies to ameliorate this dose-limiting side effect and for the development of novel safer polymyxins. The primary aim of this study was to identify the major pathways which lead to polymyxin-induced apoptosis in cultured rat kidney proximal tubular cells (NRK-52E). Caspase-3, -8, and -9 were activated by polymyxin B treatment in a concentration-dependent manner. Concentration- and time-dependent expression of FasL and deformation of mitochondrial morphology were revealed following polymyxin B treatment. The proportion of cells with filamentous mitochondria (regular morphology) following an 8-h treatment with 1.0 mM polymyxin B was 56.2% ± 9.7% (n = 3). This was decreased to 30.7% ± 7.5% when the polymyxin B concentration was increased to 2.0 mM. The mitochondrial membrane potential (Δψm) decreased to 14.1% ± 2.9% in the cells treated with 1.0 mM polymyxin B for 24 h (n = 3) compared to that in the untreated control group. Concomitantly, concentration- and time-dependent production of mitochondrial superoxide was also observed. This study is the first to have demonstrated that polymyxin-induced apoptosis is mediated through both the death receptor and mitochondrial pathways in cultured renal tubular cells. It provides key information not only for the amelioration of polymyxin-induced nephrotoxicity but also for the discovery of novel safer polymyxin-like antibiotics against Gram-negative "superbugs."
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
87917
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
C26H27ClN2O7