EMAIL THIS PAGE TO A FRIEND
Journal of clinical oncology : official journal of the American Society of Clinical Oncology
Phase II study of everolimus and letrozole in patients with recurrent endometrial carcinoma.
PMID 25624430
Abstract
The phosphoinositol-3 kinase (PI3K) pathway is frequently dysregulated in endometrial cancer (EC). Hormonal manipulation leads to response in some patients with EC, but resistance derived from PI3K pathway activation has been documented. Targeting mammalian target of rapamycin (mTOR) may overcome endocrine resistance. We conducted a two-institution phase II trial of everolimus and letrozole in women with recurrent EC. Patients were considered incurable, had measurable disease, and were treated with up to two prior cytotoxic regimens. Everolimus was administered orally at 10 mg daily and letrozole was administered orally at 2.5 mg daily. Each cycle consisted of 4 weeks of therapy. Patients were treated until progression, toxicity, or complete response (CR). The primary end point was the clinical benefit rate (CBR), which was defined as CR, partial response, or stable disease (≥ 16 weeks) by RECIST 1.0 criteria. Translational studies were performed to correlate biomarkers with response. Thirty-eight patients were enrolled (median age, 62 years; range, 24 to 82 years). Thirty-five patients were evaluable for response. The CBR was 40% (14 of 35 patients); the median number of cycles among responders was 15 (range, seven to 29 cycles). The confirmed objective response rate (RR) was 32% (11 of 35 patients; nine CRs and two partial responses; median, 15 cycles; range, eight to 29 cycles). Twenty percent of patients (seven of 35 patients) were taken off treatment after a prolonged CR and at the discretion of the treating clinician. None of the patients discontinued treatment as a result of toxicity. Serous histology was the best predictor of lack of response. Patients with endometrioid histology and CTNNB1 mutations responded well to everolimus and letrozole. Everolimus plus letrozole results in a high CBR and RR in patients with recurrent EC. Further development of this combination in recurrent endometrioid EC is under way.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L6545
Letrozole, ≥98% (HPLC)
C17H11N5