EMAIL THIS PAGE TO A FRIEND
European journal of nuclear medicine and molecular imaging
Potential diagnostic value of regional myocardial adrenergic imaging using (123)I-MIBG SPECT to identify patients with Lewy body diseases.
PMID 25626701
Abstract
The aim of this study was to determine the potential diagnostic value of regional myocardial adrenergic (123)I-metaiodobenzylguanidine (MIBG) single photon emission computed tomography (SPECT) imaging to identify patients with Lewy body diseases (LBD+). Sixty-four consecutive patients who underwent cardiac (123)I-MIBG SPECT to differentiate LBD+, including Parkinson's disease (PD) and dementia with Lewy bodies (DLB), from patients without LBD (LBD-) were retrospectively reviewed. A neurologist expert in memory disorders determined the final clinical diagnosis by using international clinical diagnostic criteria. Planar [heart to mediastinum ratio (HMR)] and (123)I-MIBG SPECT[(innervation defect score (IDS)] using the 17-segment left ventricular model (five-point scale) were obtained 4 h after the injection of (123)I-MIBG on a low-energy high-resolution (LEHR) collimator. Receiver-operating characteristic (ROC) analysis was performed to determine the optimal HMR and IDS cut-off values to discriminate LBD+ from LBD-. Of the 64 patients, 45 (70 %) were diagnosed LBD+ (DLB, n = 27; PD, n = 18) and 19 were diagnosed LBD- (5 other dementias, 14 other parkinsonisms). The HMR and IDS of LBD+ were significantly different from those of LBD- (1.30 ± 0.21 vs 1.65 ± 0.26, p < 0.001; 39 ± 28 vs 8 ± 16, p = 0.001). The optimal HMR and IDS cut-off values to discriminate LBD+ (n = 45) from LBD- (n = 19) were 1.47 and 6/68, providing a sensitivity and specificity of 82.2 and 84.2% and 86.7 and 73.7%, respectively. Regional myocardial adrenergic (123)I-MIBG imaging SPECT has a potential diagnostic value to identify LBD+.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I9890
m-Iodobenzylguanidine hemisulfate salt, ≥98% (HPLC and TLC)
C8H10IN3 · 0.5H2O4S