EMAIL THIS PAGE TO A FRIEND
Journal of clinical neurophysiology : official publication of the American Electroencephalographic Society
Consensus statement on continuous EEG in critically ill adults and children, part II: personnel, technical specifications, and clinical practice.
PMID 25626777
Abstract
Critical Care Continuous EEG (CCEEG) is a common procedure to monitor brain function in patients with altered mental status in intensive care units. There is significant variability in patient populations undergoing CCEEG and in technical specifications for CCEEG performance. The Critical Care Continuous EEG Task Force of the American Clinical Neurophysiology Society developed expert consensus recommendations on the use of CCEEG in critically ill adults and children. The consensus panel describes the qualifications and responsibilities of CCEEG personnel including neurodiagnostic technologists and interpreting physicians. The panel outlines required equipment for CCEEG, including electrodes, EEG machine and amplifier specifications, equipment for polygraphic data acquisition, EEG and video review machines, central monitoring equipment, and network, remote access, and data storage equipment. The consensus panel also describes how CCEEG should be acquired, reviewed and interpreted. The panel suggests methods for patient selection and triage; initiation of CCEEG; daily maintenance of CCEEG; electrode removal and infection control; quantitative EEG techniques; EEG and behavioral monitoring by non-physician personnel; review, interpretation, and reports; and data storage protocols. Recommended qualifications for CCEEG personnel and CCEEG technical specifications will facilitate standardization of this emerging technology.