EMAIL THIS PAGE TO A FRIEND
Prescrire international
Glycopyrronium + indacaterol. A fixed-dose combination with no advantages in COPD.
PMID 25629143
Abstract
No more effective than tiotropium combined with formoterol.