EMAIL THIS PAGE TO A FRIEND
Journal of thrombosis and haemostasis : JTH
Multimodal assessment of non-specific hemostatic agents for apixaban reversal.
PMID 25630710
Abstract
Non-specific hemostatic agents, namely activated prothrombin complex concentrate (aPCC), PCC and recombinant activated factor (F) VII (rFVIIa), can be used, off-label, to reverse the effects of FXa inhibitors in the rare cases of severe hemorrhages, as no approved specific antidote is available. We have evaluated the ability of aPCC, PCC and rFVIIa to reverse apixaban. Healthy volunteer whole blood was spiked with therapeutic or supra-therapeutic apixaban concentrations and two doses of aPCC, PCC or rFVIIa. Tests performed included a turbidimetry assay for fibrin polymerization kinetics analysis, scanning electron microscopy for fibrin network structure observation, thrombin generation assay (TGA), thromboelastometry, prothrombin time and activated partial thromboplastin time. aPCC generated a dense clot constituting thin and branched fibers similar to those of a control without apixaban, increased fibrin polymerization velocity and improved quantitative (endogenous thrombin potential and peak height) as well as latency (clotting and lag times) parameters. Adding PCC also improved the fibrin and increased quantitative parameters, but fibrin polymerization kinetics and latency parameters were not corrected. Finally, rFVIIa improved latency parameters but failed to restore the fibrin network structure, fibrin polymerization velocity and quantitative parameters. aPCC was more effective than PCC or rFVIIa in reversing in vitro the effects of apixaban. aPCC rapidly triggered the development of an apparently normal fibrin network and corrected latency and quantitative parameters, whereas PCC or rFVIIa had only a partial effect.