EMAIL THIS PAGE TO A FRIEND
Journal of cellular and molecular medicine
MicroRNA-30d and microRNA-181a regulate HOXA11 expression in the uterosacral ligaments and are overexpressed in pelvic organ prolapse.
PMID 25630974
Abstract
The balanced turnover of collagen is necessary to maintain the mechanical strength of pelvic supportive connective tissues. Homeobox (HOX) A11 is a key transcriptional factor that controls collagen metabolism and homoeostasis in the uterosacral ligaments (USLs), and the deficient HOXA11 signalling may contribute to alterations in the biochemical strength of the USLs, leading to pelvic organ prolapse (POP). However, it is unknown how HOXA11 transcripts are regulated in the USLs. In this study, we found that microRNA (miRNA)-30d and 181a were overexpressed in women with POP, and their expression was inversely correlated with HOXA11 mRNA levels. The overexpression of miR-30d or 181a suppressed HOXA11 mRNA and protein levels in 293T cells, whereas the knockdown of these miRNAs enhanced HOXA11 levels and collagen production. Cotransfection of a luciferase reporter plasmid containing the 3'-untranslated region of HOXA11 with miR-30d or 181a mimic resulted in decreased relative luciferase activity. Conversely, cotransfection with anti-miR-30d or 181a increased luciferase activity. Taken together, these results indicate that both miR-30d and 181a are important posttranscriptional regulators of HOXA11 in the USLs and could be a potential therapeutic target for POP.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN10230 CompoZr® Knockout ZFN Kit, Human HOXA11 (NM_005523)