EMAIL THIS PAGE TO A FRIEND
Journal of clinical microbiology
Development and validation of a rapid immunochromatographic assay for detection of Middle East respiratory syndrome coronavirus antigen in dromedary camels.
PMID 25631809
Abstract
We present here a rapid immunochromatographic assay for the detection of Middle East respiratory syndrome coronavirus (MERS-CoV) antigen in the nasal swabs of dromedary camels. The assay is based on the detection of MERS-CoV nucleocapsid protein in a short time frame using highly selective monoclonal antibodies at room temperature. The relative sensitivity and specificity of the assay were found to be 93.90% and 100%, respectively, compared to that of the UpE and open reading frame 1A (Orf1A) real-time reverse transcriptase PCR (RT-PCR). The results suggest that the assay developed here is a useful tool for the rapid diagnosis and epidemiological surveillance of MERS-CoV infection in dromedary camels.
Related Materials