EMAIL THIS PAGE TO A FRIEND
ZooKeys
Integrative taxonomy reveals a new species of Callisto (Lepidoptera, Gracillariidae) in the Alps.
PMID 25632257
Abstract
Europe has one of the best-known Lepidopteran faunas in the world, yet many species are still being discovered, especially in groups of small moths. Here we describe a new gracillariid species from the south-eastern Alps, Callistobasistrigella Huemer, Deutsch & Triberti, sp. n. It shows differences from its sister species Callistocoffeella in morphology, the barcode region of the cytochrome c oxidase I gene and the nuclear gene histone H3. Both Callistobasistrigella and Callistocoffeella can co-occur in sympatry without evidence of admixture. Two Callistobasistrigella specimens show evidence of introgression. We highlight the importance of an integrative approach to delimit species, combining morphological and ecological data with mitochondrial and nuclear sequence data. Furthermore, in connection with this study, Ornixblandella Müller-Rutz, 1920, syn. n. is synonymized with Callistocoffeella (Zetterstedt, 1839).