EMAIL THIS PAGE TO A FRIEND
Diabetologia
A cautionary tale: the non-causal association between type 2 diabetes risk SNP, rs7756992, and levels of non-coding RNA, CDKAL1-v1.
PMID 25634229
Abstract
Intronic single nucleotide polymorphisms (SNPs) in the CDKAL1 gene are associated with risk of developing type 2 diabetes. A strong correlation between risk alleles and lower levels of the non-coding RNA, CDKAL1-v1, has recently been reported in whole blood extracted from Japanese individuals. We sought to replicate this association in two independent cohorts: one using whole blood from white UK-resident individuals, and one using a collection of human pancreatic islets, a more relevant tissue type to study with respect to the aetiology of diabetes. Levels of CDKAL1-v1 were measured by real-time PCR using RNA extracted from human whole blood (n = 70) and human pancreatic islets (n = 48). Expression with respect to genotype was then determined. In a simple linear regression model, expression of CDKAL1-v1 was associated with the lead type 2 diabetes-associated SNP, rs7756992, in whole blood and islets. However, these associations were abolished or substantially reduced in multiple regression models taking into account rs9366357 genotype: a moderately linked SNP explaining a much larger amount of the variation in CDKAL1-v1 levels, but not strongly associated with risk of type 2 diabetes. Contrary to previous findings, we provide evidence against a role for dysregulated expression of CDKAL1-v1 in mediating the association between intronic SNPs in CDKAL1 and susceptibility to type 2 diabetes. The results of this study illustrate how caution should be exercised when inferring causality from an association between disease-risk genotype and non-coding RNA expression.