EMAIL THIS PAGE TO A FRIEND
Chinese medical journal
Evaluation of eight different clinical staging systems associated with overall survival of chinese patients with hepatocellular carcinoma.
PMID 25635426
Abstract
Hepatocellular carcinoma (HCC) is a common cancer in China, an area of high hepatitis B virus (HBV) infection. Although several staging systems are available, there is no consensus on the best classification to use because multiple factors, such as etiology, clinical treatment and populations could affect the survival of HCC patients. This study analyzed 743 HBV-related Chinese HCC patients who received surgery first and evaluated the predictive values of eight different commonly used staging systems in the clinic. The overall 1-, 3-, 5-year survival rates and a median survival were 91.5%, 70.3%, 55.3% and 72 months respectively. Barcelona Clinic Liver Cancer (BCLC) staging systems had the best stratification ability and showed the lowest Akaike information criterion (AIC) values (2896.577), followed by tumor-node-metastasis 7 th (TNM 7 th ) (AIC = 2899.980), TNM 6 th (AIC = 2902.17), Japan integrated staging score (AIC = 2918.085), Tokyo (AIC = 2938.822), Cancer of the Liver Italian Program score (AIC = 2941.950), Chinese University Prognostic Index grade (AIC = 2962.027), and Okuda (AIC = 2979.389). BCLC staging system is a better staging model for HBV infection patients with HCC in Chinese population among the eight currently used staging systems. These identifications afford a large group of Chinese HCC patients with HBV infection and could be helpful to design a new staging system for a certain population.