EMAIL THIS PAGE TO A FRIEND
Journal of cardiovascular pharmacology
Autocrine control of angiogenesis by endogenous acetylcholine in an in vitro model using human endothelial cells: evidence for an autocrine cholinergic system in endothelial cells.
PMID 25636069
Abstract
We wanted to elucidate whether acetylcholine as the endogenous ligand at cholinoceptors (ChRs) may have effects on angiogenesis and whether they are transduced through muscarinic or nicotinic ChRs. Human umbilical vein endothelial cells were cultured until confluence and thereafter seeded in Matrigel in vitro angiogenesis assays for 18 hours. During the entire cell culture and angiogenesis period, cells were treated with vehicle, eserine (1 μM), in the absence or presence of additional atropine (1 μM) or mecamylamine (1 μM). Finally, the resulting angiogenetic network was investigated histologically. Eserine significantly enhanced acetylcholine formation. When acetylcholine acted through muscarinic ChRs (eserine + mecamylamine), we observed enhanced complexity of the angiogenic network pattern with increased tube length and cell number. In contrast, when acting through nicotinic ChRs (eserine + atropine), we found reduced complexity of pattern with less branches, shorter tubes, and reduced cell number. If acting on both types of ChRs (eserine alone), there were only very small effects. Using α-bungarotoxin, lobeline, and dihydro-β-erythroidine, we also could show that these effects to various degrees involve α7, α3/β2, and α4/β2 n-ChRs. In conclusion, our results support the hypothesis that human umbilical vein endothelial cells possess an autocrine nonneuronal cholinergic system regulating angiogenesic branch formation through the partially opposing effects of n-ChRs and m-ChRs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E8375
Eserine
C15H21N3O2
H108
Hemicholinium-3, solid, ≥95% (HPLC)
C24H34Br2N2O4