EMAIL THIS PAGE TO A FRIEND
Molecular cancer research : MCR
mTOR/MYC Axis Regulates O-GlcNAc Transferase Expression and O-GlcNAcylation in Breast Cancer.
PMID 25636967
Abstract
Cancers exhibit altered metabolism characterized by increased glucose and glutamine uptake. The hexosamine biosynthetic pathway (HBP) uses glucose and glutamine, and directly contributes to O-linked-β-N-acetylglucosamine (O-GlcNAc) modifications on intracellular proteins. Multiple tumor types contain elevated total O-GlcNAcylation, in part, by increasing O-GlcNAc transferase (OGT) levels, the enzyme that catalyzes this modification. Although cancer cells require OGT for oncogenesis, it is not clear how tumor cells regulate OGT expression and O-GlcNAcylation. Here, it is shown that the PI3K-mTOR-MYC signaling pathway is required for elevation of OGT and O-GlcNAcylation in breast cancer cells. Treatment with PI3K and mTOR inhibitors reduced OGT protein expression and decreased levels of overall O-GlcNAcylation. In addition, both AKT and mTOR activation is sufficient to elevate OGT/O-GlcNAcylation. Downstream of mTOR, the oncogenic transcription factor c-MYC is required and sufficient for increased OGT protein expression in an RNA-independent manner and c-MYC regulation of OGT mechanistically requires the expression of c-MYC transcriptional target HSP90A. Finally, mammary tumor epithelial cells derived from MMTV-c-myc transgenic mice contain elevated OGT and O-GlcNAcylation and OGT inhibition in this model induces apoptosis. Thus, OGT and O-GlcNAcylation levels are elevated via activation of an mTOR/MYC cascade. Evidence indicates OGT as a therapeutic target in c-MYC-amplified cancers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300531
Anti-BCL2L11 (Ab-69) antibody produced in rabbit, affinity isolated antibody
PRS2065
Anti-Bim (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3405
Anti-Bim (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
B7929
Anti-Bim antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1405706
Anti-DDIT3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
G6916 Anti-GADD153 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500632
Anti-GADD153 antibody produced in rabbit, affinity isolated antibody
SAB1407043
Anti-MGEA5 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA036141
Anti-MGEA5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4200267
Anti-O-GlcNAcase (OGA) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody
SAB2500715
Anti-OGT antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2101676
Anti-OGT antibody produced in rabbit, affinity isolated antibody
HPA030751
Anti-OGT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA030752
Anti-OGT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
PRS3507
Anti-TSC2 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3509
Anti-TSC2 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA030409
Anti-TSC2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503037
Anti-TSC2 antibody produced in rabbit, affinity isolated antibody
SAB4503038
Anti-TSC2, C-Terminal antibody produced in rabbit, affinity isolated antibody
T9574
Anti-Tuberin (IA-22) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
T5075 Anti-Tuberin (VV-18) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503039
Anti-Tuberin antibody produced in rabbit, affinity isolated antibody
WH0010018M1 Monoclonal Anti-BCL2L11 antibody produced in mouse, clone 2F10, purified immunoglobulin, buffered aqueous solution
WH0001649M1
Monoclonal Anti-DDIT3 antibody produced in mouse, clone 2G3, purified immunoglobulin, buffered aqueous solution
SAB1403729
Monoclonal Anti-DDIT3 antibody produced in mouse, clone 2C4, purified immunoglobulin, buffered aqueous solution
SAB1403082
Monoclonal Anti-MGEA5, (C-terminal) antibody produced in mouse, clone 1C7, purified immunoglobulin, buffered aqueous solution
SAB1402949
Monoclonal Anti-TSC2 antibody produced in mouse, clone 1C1, purified immunoglobulin, buffered aqueous solution