EMAIL THIS PAGE TO A FRIEND
The Journal of infectious diseases
The Intrahepatic T Cell Compartment Does Not Normalize Years After Therapy-Induced Hepatitis C Virus Eradication.
PMID 25637348
Abstract
Little is known about the immune status in liver and blood of patients with chronic hepatitis C virus (HCV) infection long after therapy-induced viral clearance. In this study, we demonstrate that, 4 years after clearance, regulation of HCV-specific immunity in blood by regulatory T cells (Tregs) and the immunosuppressive cytokines interleukin 10 and transforming growth factor β is still ongoing. Importantly, analysis of liver specimens collected 4 years after HCV clearance shows that intrahepatic Tregs are still present in all patients, suggesting that liver T cells remain regulated. Identifying mechanisms that regulate HCV-specific memory T-cell responses after clearance is highly relevant for the development of protective vaccines, especially in patients at high risk of reinfection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R9644
Ribavirin, antiviral
C8H12N4O5