EMAIL THIS PAGE TO A FRIEND
Fertility and sterility
Insulin resistance and hyperandrogenism have no substantive association with birth weight in adolescents with polycystic ovary syndrome.
PMID 25637475
Abstract
To assess whether birth weight influences the metabolic and hormonal profile of adolescents with polycystic ovary syndrome (PCOS). Retrospective study. University outpatient clinic. One hundred seventy consecutive adolescents 12 to 19xa0years of age with PCOS, 15 of whom were small for gestational age (SGA), and 75 healthy female aged-matched adolescents as controls. Physical evaluations, fasting blood samples for measuring endocrine and metabolic parameters, and an oral glucose tolerance test. Physical, endocrine, and metabolic features. The birth weights of adolescents with PCOS as well as those with hyperinsulinemic or insulin resistance were similar to those of the control group. The PCOS SGA adolescents had basal insulin (15.93 ± 7.16 μU/mL vs. 10.97 ± 5.79 μU/mL) and homeostasis model assessment of insulin resistance values (3.2 ± 1.54 vs. 2.19 ± 1.28) that were statistically significantly higher than in the control group. The mean levels of total testosterone in the SGA adolescents with PCOS were above the upper limit of the normal range (0.80xa0ng/mL). Low birth weight may influence the appearance of hyperandrogenism and insulin resistance in a portion of adolescents with PCOS, but only 9% of the adolescents with PCOS in this study were SGA. In the majority of adolescents with PCOS, hyperinsulinemia and hyperandrogenism are related to factors other than birth weight alone.