EMAIL THIS PAGE TO A FRIEND
Steroids
3,16-Bisquaternary ammonium steroid derivatives as neuromuscular blocking agents: synthesis and biological evaluation.
PMID 25637675
Abstract
Neuromuscular blocking agents (NMBAs) are widely used in surgery to achieve skeleton muscles relaxation under light anesthesia status. In this work, we synthesized a series of 3,16-bisquaternary ammonium steroidal NMBAs. Among them, three compounds exhibited higher in vitro activities than the commenced drug rocuronium. In addition, structure-activity relationship was unveiled. We found that the intact acetylcholine-like moiety in D-ring was not necessary for maintaining activity but both the acetyl group and the quaternary nitrogen were very essential.