EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis of (+)-crocacin D and simplified bioactive analogues.
PMID 25638500
Abstract
The total synthesis of (+)-crocacin D has been achieved in 15 steps (9 isolated intermediates) and 14% overall yield from commercially available starting materials and using (+)-crocacin C as a key intermediate. A number of simplified analogues and their biological activities are also reported.