EMAIL THIS PAGE TO A FRIEND
Food and nutrition bulletin
Assessing the potential of wild foods to reduce the cost of a nutritionally adequate diet: an example from eastern Baringo District, Kenya.
PMID 25639131
Abstract
Wild foods and their actual and potential contributions to nutrition security have rarely been studied or considered in nutrition and conservation programs. To study the role of wild food biodiversity in achieving a cost reduction of a nutritionally adequate diet for women and young children in Kenya using linear programming. An ethnobiological inventory of available food biodiversity was carried out by means of focus group discussions, and five wild foods were selected for further modeling. A market survey assessed available food prices by season. Diets were modeled to minimize cost and maximize nutrient adequacy using the Cost of Diet linear programming tool. Modeling was done without and with wild foods. The modeled diets without wild species were deficient in iron for all age groups during the dry season, deficient in vitamin B6 and calcium for infants aged 6 to 8 months during the dry season, and deficient in iron and zinc for infants aged 6 to 8 months over the whole year. Adding wild foods, especially Berchemia discolor, to the modeled diets resulted in a lower-cost diet, while meeting recommended iron intakes for women and children between 12 and 23 months of age. Even after integrating wild foods into the model, targeted approaches are needed to meet micronutrient requirements for infants from 6 to 8 and from 9 to 11 months of age. An application of linear programming to screen available wild foods for meeting recommended nutrient intakes at a minimal cost was illustrated. This type of study helps to objectively assess the potential of biodiversity to contribute to diets and nutrition.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3518
Carbonyl iron, ≥97% Fe basis
Fe
44890
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Fe
12310
Iron, ≥99%, reduced, powder (fine)
Fe
267945
Iron, chips, 99.98% trace metals basis
Fe
338141
Iron, foil, thickness 0.25 mm, ≥99.99% trace metals basis
Fe
356808
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Fe
413054
Iron, granular, 10-40 mesh, >99.99% trace metals basis
Fe
513423
Iron, activated, nanoparticles, dispersion, <100 nm particle size, in mineral oil, 99.5% trace metals basis
Fe
IRMM524A
Iron, IRMM® certified Reference Material, 0.1 mm foil
Fe
IRMM524B
Iron, IRMM® certified Reference Material, 0.5 mm wire
Fe
255637
Iron, powder, ≥99.99% trace metals basis
Fe
209309
Iron, powder, −325 mesh, 97%
Fe
267953
Iron, powder, <10 μm, ≥99.9% trace metals basis
Fe
266213
Iron, rod, diam. 6.3 mm, 99.98% trace metals basis
Fe
266256
Iron, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Fe
266248
Iron, wire, diam. 0.5 mm, ≥99.99%
Fe
356824
Iron, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Fe
356832
Iron, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Fe
GF58945078
Iron, wire reel, 250m, diameter 0.25mm, hard, 99.5%
Fe
GF21695722
Iron, wire reel, 2m, diameter 0.13mm, hard, 99.99+%
Fe
GF81282135
Iron, wire reel, 2m, diameter 0.25mm, annealed, 99.99+%
Fe
GF57565996
Iron, wire reel, 2m, diameter 0.25mm, hard, 99.99+%
Fe
GF48022152
Iron, wire reel, 2m, diameter 0.5mm, hard, 99.99+%
Fe
GF48020947
Iron, wire reel, 500m, diameter 0.125mm, hard, 99.5%
Fe
GF58809965
Iron, wire reel, 50m, diameter 0.065mm, hard, 99.5%
Fe
GF34071566
Iron, wire reel, 50m, diameter 0.25mm, hard, 99.5%
Fe
GF96915134
Iron, wire reel, 50m, diameter 0.5mm, as drawn, 99.95%
Fe
GF35663935
Iron, wire reel, 5m, diameter 0.025mm, as drawn, 99.99+%
Fe
GF27511781
Iron, wire reel, 5m, diameter 0.25mm, annealed, 99.99+%
Fe
GF61040023
Iron, wire reel, 5m, diameter 0.25mm, hard, 99.99+%
Fe