EMAIL THIS PAGE TO A FRIEND
Journal of neuroscience research
Reductions in kinesin expression are associated with nitric oxide-induced axonal damage.
PMID 25639260
Abstract
Axonal injury is often characterized by axonal transport defects and abnormal accumulation of intra-axonal components. Nitric oxide (NO) has a key role in mediating inflammatory axonopathy in many neurodegenerative diseases, but little is known about how nitrosative/oxidative stress affects axonal transport or whether reductions in kinesin superfamily protein (KIF) expression correlate with axon pathology. KIFs are molecular motors that have a key role in axonal and dendritic transport, and impairment of these mechanisms has been associated with a number of neurological disorders. This study shows that rat cortical neurons exposed to NO display both a time-dependent decrease in KIF gene/protein expression and neurofilament phosphorylation in addition to a reduction in axonal length and neuronal survival. Because mesenchymal stem cells (MSCs) represent a promising therapeutic candidate for neuronal/axonal repair, this study analyzes the capacity of MSCs to protect neurons and axonal transport mechanisms from NO damage. Results show that coculture of MSCs with NO-exposed neurons results in the preservation of KIF expression, axonal length, and neuronal survival. Altogether, these results suggest a potential mechanism involved in the disruption of axonal transport and abnormal accumulation of proteins in axons during nitrosative insult. We hypothesize that impaired axonal transport contributes, per se, to progression of injury and provide further evidence of the therapeutic potential of MSCs for neurodegenerative disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA010926
Anti-ALCAM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1306488
ANTI-CD166(N-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2701669
Anti-eno2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1408571
Anti-ENO2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA027274
Anti-KIF21B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA027249
Anti-KIF21B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA004469
Anti-KIF5A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV33868
Anti-KIF5A antibody produced in rabbit, affinity isolated antibody
SAB1305394
ANTI-NSE (Y236) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1305395
ANTI-NSE (Y25) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300698
Anti-NSE antibody produced in rabbit, affinity isolated antibody
SAB4500768
Anti-NSE, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1402685
Monoclonal Anti-ALCAM antibody produced in mouse, clone 1H3, ascites fluid
SAB5300346
Monoclonal Anti-ALCAM antibody produced in mouse, clone 3F8B12, ascites fluid
SAB5300347 Monoclonal Anti-ALCAM antibody produced in mouse, clone 10F1G12, ascites fluid
WH0004744M1
Monoclonal Anti-NEFH antibody produced in mouse, clone 2E8, purified immunoglobulin, buffered aqueous solution